Learn More
VCAM-1 Monoclonal Antibody (429 (MVCAM.A)), Invitrogen™
Rat Monoclonal Antibody
Supplier:  Thermo Scientific MA546200
Encompass
Catalog No. PIMA546200
Add to Cart
This item is not returnable. View return policy
Description
Description
VCAM-1 Monoclonal antibody specifically detects VCAM-1 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VCAM-1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
P29533
VCAM1
Murine preadipose cell line PA6
100 μg
Primary
Mouse
Liquid
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
429 (MVCAM.A)
Unconjugated
VCAM1
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B
Rat
Protein G
RUO
22329
4° C, do not freeze
IgG2a κ
Recommendations
Recommendations