Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM1 Mouse anti-Human, Clone: VCAM1/843, Abnova™

Mouse monoclonal antibody raised against full length recombinant human VCAM1.

Manufacturer:  Abnova Corporation MAB13449

Catalog No. 89-935-085


Add to cart

Description

Description

This gene is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms have been described for this gene. [provided by RefSeq]

Specifications

Specifications

VCAM1
VCAM1/843
Mouse monoclonal antibody raised against full length recombinant human VCAM1.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CD106/DKFZp779G2333/INCAM-100/MGC99561
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
VCAM1
Recombinant protein corresponding to full length human VCAM1.
Protein A/G purification
RUO
Primary
7412
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit