Learn More
Electron Microscopy SciencesSupplier Diversity Partner Enlarger VCCE Dichroic Module
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 7487220
Encompass_Preferred
Interchangeable slide-in filtration modules, fits all Saunders/LPL 4550XLG and 4500-II enlargers, both models are easily converted by switching the modules.
Catalog No. 50-250-80