Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCP, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300588AM

 View more versions of this product

Catalog No. 50-155-7983


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict

The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.
Specifications

Specifications

VCP
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
P55072, Q01853
Valosin-Containing Protein, p97
Rabbit
IgG
100 μl
4° C
Primary
269523, 7415
Immunohistochemistry, Western Blot
Unconjugated
VCP
liquid
Vcp
Between 1 and 50
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit