Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC2 Mouse anti-Human, Mouse, Rat, Clone: 1D8C11, Proteintech Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 663881IG

Catalog No. 50-114-2818


Add to cart

Description

Description

This gene encodes a member of the voltage-dependent anion channel pore-forming family of proteins that are considered the main pathway for metabolite diffusion across the mitochondrial outer membrane. The encoded protein is also thought to be involved in the mitochondrial apoptotic pathway via regulation of BCL2-antagonist/killer 1 protein activity. Pseudogenes have been identified on chromosomes 1, 2, 12 and 21, and alternative splicing results in multiple transcript variants.
Specifications

Specifications

VDAC2
1D8C11
Unconjugated
VDAC2
Liquid
VDAC2
Peptide
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P45880, P81155, Q60930
hVDAC2, VDAC 2, VDAC2
Mouse
IgG1
150 μL
-20°C
Primary
22334, 7417, 83531
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit