Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC2 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP220849AF350

Catalog No. NP220849A30


Add to cart

Description

Description

VDAC2 Polyclonal antibody specifically detects VDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

VDAC2
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
VDAC2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VDAC2. The exact sequence is proprietary.
Immunogen affinity purified
Lipid and Metabolism
Primary
7417.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit