Learn More
VDAC2 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 700, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP220849AF700
Catalog No. NP220849A70
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
VDAC2 Polyclonal antibody specifically detects VDAC2 in Human,Mouse,Rat samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
VDAC2
Alexa Fluor™ 700
50 mM sodium borate with 0.05% sodium azide
VDAC2
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VDAC2. The exact sequence is proprietary.
0.1 mL
Primary
7417
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2
Rabbit
IgG
Store at 4°C in the dark.
Polyclonal
Human, Mouse, Rat