Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDAC3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551156
Description
The antibody detects endogenous levels of total VDAC3 protein.
VDAC3 Polyclonal antibody specifically detects VDAC3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
VDAC3 | |
Polyclonal | |
Unconjugated | |
VDAC3 | |
channel protein VDAC3; HD-VDAC3; mitochondrial outer membrane protein; mitochondrial voltage dependent anion channel 3; mVDAC3; outer mitochondrial membrane protein porin 3; rVDAC3; Vdac3; VDAC-3; voltage dependent anion channel 3; voltage-dependent anion channel 3; voltage-dependent anion-selective channel protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7419 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y277 | |
VDAC3 | |
Full length fusion protein of human VDAC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction