VDM 11 (in Tocrisolve 100), Tocris

Potent, selective anandamide transport inhibitor (in water-soluble emulsion)

Manufacturer: R&D Systems 1686/25

Quantity 25mg

 View more versions of this product

Catalog No. 16-862-5

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C27H39NO2
    Formula Weight 409.61
    Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
    Chemical Name or Material VDM 11 (in Tocrisolve™ 100)

    Specifications

    Formula Weight 409.61
    Quantity 25mg
    Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
    Assay Percent Range >98%
    Molecular Formula C27H39NO2
    CAS 313998-81-1
    Recommended Storage Store at +4°C
    Chemical Name or Material VDM 11 (in Tocrisolve™ 100)

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit