VDM 11, Tocris

Potent, selective anandamide transport inhibitor

Manufacturer: R&D Systems 1392/5

Quantity 5mg

 View more versions of this product

Catalog No. 13-925

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C27H39NO2
    Formula Weight 409.61
    Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
    Chemical Name or Material VDM 11

    Specifications

    Formula Weight 409.61
    Quantity 5mg
    Synonym (5Z,8Z,11Z,14Z)-N-(4-Hydroxy-2-methylphenyl)-5,8,11,14-eicosatetraenamide
    Assay Percent Range >98%
    Molecular Formula C27H39NO2
    CAS 313998-81-1
    Recommended Storage Desiccate at -20°C
    Chemical Name or Material VDM 11

    Desiccate at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit