Learn More
Cayman Chemical VDM11 Fatty Acids and Derivati
SDP
Supplier:  Cayman Chemical 100067315 MG
Encompass_Preferred
An AEA transport inhibitor with essentially no activity on the CB1 receptor, CB2 receptor, or TRPV1; inhibits FAAH and MAGL and may act as an alternative FAAH substrate; inhibits glutamergic synaptic transmission between hippocampal neurons at a concentration of 3 µM
Catalog No. 50-289-0832