Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VE-Cadherin Mouse anti-Human, Alexa Fluor 594, Clone: 123413, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB9381T100UG
This item is not returnable.
View return policy
Description
VE-Cadherin Monoclonal specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry.Specifications
VE-Cadherin | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P33151 | |
CDH5 | |
Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin Asp48-Gln593 Accession # P33151 | |
RUO | |
1003 | |
Human | |
IgG2b |
Flow Cytometry | |
123413 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin | |
Mouse | |
100 μg | |
Primary | |
Detects human VE-Cadherin in Western blots. In Western blots, 25% cross-reactivity with recombinant mouse VE-Cadherin and no cross-reactivity with recombinant human (rh) Cadherin-17 or rhP-Cadherin is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)