Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Cadherin Mouse anti-Human, Alexa Fluor 700, Clone: 123413, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB9381N100UG

Catalog No. FAB9381N100


Add to cart
This item is not returnable. View return policy

Description

Description

VE-Cadherin Monoclonal antibody specifically detects VE-Cadherin in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

VE-Cadherin
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P33151
CDH5
Mouse myeloma cell line NS0-derived recombinant human VE-Cadherin Asp48-Gln593 Accession # P33151
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human VE-Cadherin in Western blots. In Western blots, 25% cross-reactivity with recombinant mouse VE-Cadherin and no cross-reactivity with recombinant human (rh) Cadherin-17 or rhP-Cadherin is observed.
Flow Cytometry
123413
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, CDH5, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin, VECadherin
Mouse
IgG2b
RUO
Primary
1003
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)