Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VE-Cadherin Mouse anti-Human, Rat, PE, Clone: 3D5C7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237420PE

Catalog No. NBP237420PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

VE-Cadherin
3D5C7
Flow Cytometry
Purified
CDH5
Purified recombinant fragment of human CDH5 (AA: 29-223 ) expressed in E. Coli.
Protein A or G purified
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Phospho Specific, Plasma Membrane Markers, Signal Transduction
Primary
1003
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
7B4, 7B4 antigen, cadherin 5, type 2 (vascular endothelium), cadherin 5, type 2, VE-cadherin (vascular epithelium), cadherin-5, CD144, CD144 antigen, endothelial-specific cadherin, FLJ17376, Vascular endothelial cadherin, VE-cadherin
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit