Learn More
Novus Biologicals VECTASTAIN(R) ABC anti-mouse/rabbit AP Immunodetection Kit
Novus Biologicals VECTASTAIN(R) ABC anti-mouse/rabbit AP Immunodetection Kit is an alkaline phosphatase enzyme detection system.
Manufacturer: Novus Biologicals AK5200NB
Description
- Avidin/biotin technology
- High sensitivity/low background
- Preferred for tissues that have high endogenous peroxidase activity
- Offers additional substrate choices
- Used as a second label in multiple antigen labeling applications
- Kit contents:
- 2mL Reagent B (Biotinylated AP)
- 2mL Reagent A (Avidin DH)
- 2mL Biotinylated Horse Anti-Mouse/Rabbit IgG
- 3mL Normal Horse Serum
Specifications
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, In-situ Hybridization, ELISPOT | |
1 Kit | |
Store at 4C. Do not freeze. | |
AP Immunodetection Kit |