Learn More
Cell Biolabs IncSupplier Diversity Partner VECTOR EXP PAAV-IRES-GFP  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK418
Clone your gene into the pAAV-IRES-GFP Expression Vector and then co-transfect with packaging plasmids to generate recombinant AAV.
Catalog No. 50-673-875