Learn More
Cell Biolabs IncSupplier Diversity Partner VECTOR EXP PSCAAV-MCS  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner VPK430
Clone your gene into the pscAAV-MCS Expression Vector and then co-transfect with packaging plasmids to generate recombinant self-complementary AAV.
Catalog No. 50-673-901