Learn More
Cell Biolabs IncSupplier Diversity Partner VECTOR RETRO CLON PMXS-PURO  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner RTV012
Clone your gene of interest into the pMXs-Puro Retroviral Vector and then package into a retroviral particle using a retroviral packaging cell or co-transfect with retroviral packaging constructs into 293T cells.
Catalog No. 50-673-340