Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF 121 Rabbit anti-Human, HRP, Clone: 64, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP289193H
Description
VEGF 121 Monoclonal antibody specifically detects VEGF 121 in Human samples. It is validated for ELISA, ELISASpecifications
VEGF 121 | |
64 | |
ELISA, Sandwich ELISA Detection | |
Purified | |
Rabbit | |
IgG | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins | |
Primary | |
7422 |
ELISA | |
HRP | |
PBS | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Recombinant Human VEGF 121 protein | |
Protein A purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title