Learn More
VEGF-B 167/186 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF590
 View more versions of this product
Catalog No. AF590
Add to cart
This item is not returnable. View return policy
Description
Description
VEGF-B 167/186 Polyclonal antibody AF590 Specifically detects VEGF-B 167/186 in Mouse samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications
Specifications
VEGF-B
LYOPH
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction
P49766
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects mouse VEGF-B 167 and mouse VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with rhVEGF-B 186 is observed, 15% cross-reactivity with rhVEGF-B 167 is observed and less than 1% cross-reactivity with rmVEGF 164 rhVEGF-C and rmVEGF-D is observed.
Mouse
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Vegfa
E. coli-derived recombinant mouse VEGF-B 186 Pro22-Ala107 Accession # P49766
Immunogen affinity purified
RUO
Primary
7423
Reconstitute at 0.2 mg/mL in sterile PBS.