Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-B Rat anti-Mouse, Alexa Fluor 405, Clone: RM0008-6E72, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11061024AF405
Description
VEGF-B Monoclonal specifically detects VEGF-B in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VEGF-B | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VEGFB | |
Mouse recombinant VEGF-B | |
0.1 mL | |
Primary | |
This antibody recognizes mouse VEGF-B 186 but not 167 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0008-6E72 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
vascular endothelial growth factor B, VEGF-B, VEGF-related factor, VRFVEGFL | |
Rat | |
Protein G purified | |
Angiogenesis, Cell Cycle and Replication | |
7423 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title