Learn More
Novus Biologicals™ VEGF-C ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192698
Catalog No. NBP192698
Add to cart
Description
Description
The Rat VEGF-C ELISA Kit (Colorimetric) measures rat VEGF-C in cell culture supernatant, serum, plasma (heparin).
Specifications
Specifications
47.0 - 3,000pg/mL
7424
ELISA Kit
27.0pg/mL
Storage of components varies. See protocol for specific instructions.
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
ELISA
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to rat VEGF-C, Biotin-Conjugate anti-rat VEGF-C monoclonal antibody, Streptavidin-HRP, Rat VEGF-C Standard lyophilized, 6ng/mL upon reconstitution, Sample Diluent, Conjugate Diluent, Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma (heparin)
VEGF-C
VEGFC