Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Mouse anti-Human, Biotin, Clone: MM0006-2E65, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB11060971B

Catalog No. NB11060971B


Add to cart

Description

Description

VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

VEGF-C
MM0006-2E65
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P49767
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
No cross-reactivity is shown with human VEGFA, VEGF-B and VEGF-D.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Biotin
PBS with 0.05% Sodium Azide
Purified
VEGFC
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with recombinant human VEGF-C recombinant protein.
Protein G purified
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary
7424
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit