Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF-C Mouse anti-Human, DyLight™ 350, Clone: OTI4A1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274845UV

Catalog No. NBP274845UV


Add to cart

Description

Description

VEGF-C Monoclonal antibody specifically detects VEGF-C in Human samples. It is validated for Western Blot.
Specifications

Specifications

VEGF-C
OTI4A1
Western Blot
Affinity Purified, Conjugated
VEGFC
Recombinant protein fragment corresponding to amino acids 112-227 of human VEGF-C (NP_005420) produced in E.coli.
Immunogen affinity purified
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary
7424.0
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit