Learn More
VEGF-C Mouse anti-Human, DyLight 550, Clone: MM0006-2E65, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11060971R
Catalog No. NB11060971R
Add to cart
Description
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
VEGF-C
MM0006-2E65
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P49767
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
No cross-reactivity is shown with human VEGFA, VEGF-B and VEGF-D.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Purified
VEGFC
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with recombinant human VEGF-C recombinant protein.
Protein G purified
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia
Primary
7424
Human