Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Antibody (9/G10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP236287AF750
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
VEGF-C | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
VEGFC | |
Recombinant human VEGF-C | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Western Blot, ELISA | |
9/G10 | |
Western Blot, ELISA | |
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
Mouse | |
Protein G purified | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
7424.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction