Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-C Mouse anti-Human, Rat, Clone: 107/A11, Novus Biologicals™
Mouse Monoclonal Antibody
$212.24 - $493.00
Specifications
Antigen | VEGF-C |
---|---|
Clone | 107/A11 |
Concentration | LYOPH |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Description
VEGF-C Monoclonal specifically detects VEGF-C in Human, Rat samples. It is validated for Western Blot, ELISA, Sandwich ELISA.Specifications
VEGF-C | |
LYOPH | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer, Cell Biology, HIF Target Genes, Hypoxia | |
Flt4 ligand, Flt4-L, vascular endothelial growth factor C, Vascular endothelial growth factor-related protein, VEGF-C, VRPFLT4 ligand DHM | |
VEGFC | |
IgG2a | |
Protein G purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
107/A11 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Sandwich ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
7424 | |
Recombinant human VEGF-C | |
Primary | |
Centrifuge vial prior to opening. Reconstitute in sterile water to a concentration of 0.1-1.0 mg/ml. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title