Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF-D Rabbit anti-Human, Clone: 106, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | VEGF-D |
---|---|
Clone | 106 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
VEGF-D Monoclonal specifically detects VEGF-D in Human samples. It is validated for ELISA.Specifications
VEGF-D | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
c-fos induced growth factor (vascular endothelial growth factor D), vascular endothelial growth factor D, VEGF-DVEGFDc-Fos-induced growth factor | |
Recombinant Human VEGF-D protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
106 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Cellular Markers | |
0.2 um filtered solution in PBS | |
2277 | |
IgG | |
Protein A purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title