Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGF Mouse anti-Human, DyLight 755, Clone: 6D3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP118621IR
Description
VEGF Monoclonal specifically detects VEGF in Human samples. It is validated for Western Blot, ELISA.Specifications
VEGF | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VEGFA | |
recombinant human P38-delta | |
0.2 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, GPCR, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision | |
7422 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA | |
6D3 | |
Western Blot, ELISA | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title