Learn More
VEGFR3/Flt-4 Mouse anti-Human, DyLight 488, Clone: MM0003-7G63, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11060968G
Catalog No. NB11060968G
Add to cart
Description
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
VEGFR3/Flt-4
MM0003-7G63
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
FLT4
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human VEGFR3 N-terminal recombinant protein.
Protein G purified
Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary
2324
Human
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This detects human VEGFR3 in western blots. No cross-reactivity is shown with VEGFR1 or VEGFR2.