Learn More
VEGFR3/Flt-4 Rat anti-Mouse, DyLight 405, Clone: RM0003-5F63, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11061018V
Catalog No. NB11061018V
Add to cart
Description
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
VEGFR3/Flt-4
RM0003-5F63
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
FLT4
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with purified mouse VEGFR3 extracellular domain.
Protein G purified
Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary
2324
Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Rat
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
No cross reactivity to mouse VEGFR1 and VEGFR2.