Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFA Mouse anti-Human, Clone: VG-1, Abnova™

Mouse monoclonal antibody raised against human VEGFA.

Manufacturer:  Abnova Corporation MAB12579

Catalog No. 89-935-090


Add to cart

Description

Description

This gene is a member of the PDGF/VEGF growth factor family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Elevated levels of this protein is linked to POEMS syndrome, also known as Crow-Fukase syndrome. Mutations in this gene have been associated with proliferative and nonproliferative diabetic retinopathy. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. There is also evidence for the use of non-AUG (CUG) translation initiation sites upstream of, and in-frame with the first AUG, leading to additional isoforms. [provided by RefSeq]

Specifications

Specifications

VEGFA
VG-1
Mouse monoclonal antibody raised against human VEGFA.
In PBS (0.2% gelatin, 0.09% sodium azide).
MGC70609/VEGF/VEGF-A/VPF
Mouse
IgG1
50 ug
Store at 4°C. Do not freeze.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) Western Blot (1:200) The optimal working dilution should be determined by the end user.
Liquid
VEGFA
Recombinant protein corresponding to human VEGFA.
Protein A/G purification
RUO
Primary
7422
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit