Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR1/Flt-1 Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: MAB0702, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221963AF532

Catalog No. NP221963A32


Add to cart

Description

Description

VEGFR1/Flt-1 Monoclonal antibody specifically detects VEGFR1/Flt-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

VEGFR1/Flt-1
MAB0702
Western Blot
Affinity Purified, Conjugated
FLT1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified extracellular domain of recombinant human VEGFR1. VEGFR-1 ( Antagonistic) EC domain, AAC50060.
Immunogen affinity purified
Angiogenesis, Cancer, Cellular Markers, Endothelial Cell Markers, HIF Target Genes, Hypoxia, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Markers, Tumor Suppressors, Tyrosine Kinases
Primary
2321.0
Human, Mouse
Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
EC 2.7.10, EC 2.7.10.1, FLT, FLT-1, Fms-like tyrosine kinase 1, fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascularpermeability factor receptor), FRT, Tyrosine-protein kinase FRT, Tyrosine-protein kinase receptor FLT, vascular endothelial growth factor receptor 1, Vascular permeability factor receptor, VEGFR1, VEGFR-1
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes both human and mouse VEGFR1 in western blot. It shows no cross-reactivity with hVEGFR2 and mVEGFR2
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit