Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR1 Rabbit anti-Human, Non-human Primate, Biotin, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc VEGFR1BIOTIN

Description
VEGFR1 Polyclonal antibody specifically detects VEGFR1 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
VEGFR1 | |
Polyclonal | |
Biotin | |
FLT1 | |
AI323757; embryonic receptor kinase 2; Emrk2; FLT; FLT1; FLT-1; fms related tyrosine kinase 1; FMS-like tyrosine kinase 1; FMS-related tyrosine kinase 1; Fms-related tyrosine kinase 1 (vascular endothelial growth factor/vascular permeability factor receptor); FRT; i21; sFlt1; soluble VEGFR 1; sVEGFR 1; Tyrosine-protein kinase FRT; tyrosine-protein kinase receptor FLT; Vascular endothelial growth factor receptor 1; vascular endothelial growth factor receptor-1; vascular permeability factor receptor; VEGF R1; VEGFR1; VEGFR-1 | |
Synthetic peptide taken within amino acid region 1250-1300 on human Vascular endothelial growth factor receptor 1 protein. | |
200 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P17948 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2321, 452512 | |
-20°C | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title