Learn More
VEGFR2/KDR/Flk-1 Mouse anti-Human, Alexa Fluor 700, Clone: 89106R, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB3572RN100UG
Catalog No. FAB3572RN10
Add to cart
This item is not returnable. View return policy
Description
Description
VEGFR2/KDR/Flk-1 Monoclonal specifically detects VEGFR2/KDR/Flk-1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
VEGFR2/KDR/Flk-1
89106R
Flow Cytometry 0.25-1 ug/10^6 cells
P35968
CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), KRD1, Ly73, Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VEGFR2/KDR/Flk-1, Ala20-Glu764, Accession # P35968
Protein A or G purified from cell culture supernatant
RUO
Primary
3791
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100 μg
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Human