Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VEGFR3/Flt-4 Rat anti-Human, Mouse, Clone: 7A3, Novus Biologicals™
Rat Monoclonal Antibody
$212.24 - $551.00
Specifications
Antigen | VEGFR3/Flt-4 |
---|---|
Clone | 7A3 |
Concentration | LYOPH |
Dilution | Western Blot 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
VEGFR3/Flt-4 | |
LYOPH | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rat | |
Angiogenesis, Cancer, Hypoxia, Protein Kinase | |
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
FLT4 | |
IgG1 | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
7A3 | |
Western Blot 2-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
2324 | |
mouse recombinant CXCL7 | |
Primary | |
Reconstitute the antibody with 500 ul sterile PBS and the final concentration is 200 ug/ml. | |
Rat Monoclonal to VEGFR-3 |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title