Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFR3/Flt-4 Rat anti-Mouse, Reptile, DyLight™ 550, Clone: AFL4, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143259R

Catalog No. NBP143259R


Add to cart

Description

Description

VEGFR3/Flt-4 Monoclonal antibody specifically detects VEGFR3/Flt-4 in Mouse, Reptile samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

VEGFR3/Flt-4
AFL4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
FLT4
The immunogen for this antibody was VEGFR3.
Protein A or G purified
Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary
2324.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Rat
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse, Reptile
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit