Learn More
VEGFR3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 207121AP
Catalog No. 50-173-3046
Add to cart
Description
Description
The antibody is specific to FLT4.
Vascular endothelial growth factor is a key regulator of blood vessel development in embryos and angiogenesis in adult tissues. Unlike VEGF, the related VEGFC stimulates the growth of lymphatic vessels through its specificl ymphatic endothelial receptor VEGFR-3. In 1998, Dumont showed that targeted inactivation of the VEGFR-3 gene in mice resulted in defective blood vessel development in early embryos. Vasculogenesis and angiogenesis occurred, but large vessels became abnormally organized with defective lumens, leading to fluid accumulation in the pericardial cavity and cardiovascular failure at embryonic day 9.5. Thus, VEGFR3 has an essential role in the development of theembryonic cardiovascular system before the emergence of the lymphatic vessels.
Specifications
Specifications
VEGFR3
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P35916, P35917
FLT 4, FLT4, FLT41, Fms like tyrosine kinase 4, fms related tyrosine kinase 4, LMPH1A, PCL, VEGFR 3, VEGFR3
Rabbit
IgG
150 μL
-20°C
Primary
14257, 2324
Western Blot
Unconjugated
FLT4
Liquid
FLT4
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse