Learn More
Bioworld Velveteen Transfer Squares for 100mm plates, 6X6, 1 PK(s)
Supplier: Bioworld 431700191
Reusable black velveteen squares are used to cover the upper surface of the replica plater and act as the substrate for accurate transfers. 10 Units / 1 PK