Learn More
Pyramex™ Venture 3™ Safety Eyewear
Click to view available options
Lens Tint:
Amber
Blue
Clear
Gray
IR/UV 3.0
IR/UV 5.0
Indoor/Outdoor, Mirror
Silver, Mirror
Description
Specifications
Lens Type | H2X |
Certifications/Compliance | ANSI Z87.1 |
Lens Tint | IR/UV 3.0 |
Frame Color | Black |
Weight (Metric) | 28 g |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |