Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear - Monteagle™ Safety Glasses

Venture Gear - Monteagle™ Safety Glasses with ANSI Z87.1 safety compliance

Manufacturer:  Pyramex™ VGSGM1665T

 View more versions of this product

Catalog No. 19-555-725


This item is not returnable. View return policy

Description

Description

  • Co-injected temples for snug, secure fit
  • Adjustable rubber nosepiece
  • Ice Blue Mirror Lens - Gray polycarbonate lens with a double layer of silver and blue mirror coatings, commonly used in outdoor applications
  • Lens Technology:
    • Visible Light Transmitted - 13%
    • UVA, UVB Blocked - 99%
    • Blue Light Blocked - 95%
    • IR Blocked - 51%
Specifications

Specifications

Polycarbonate
Anti-fog
Co-Injected, Polycarbonate
Eye Safety
Gun Metal
138 mm
Polycarbonate
Light Blue, Mirror
ASNI Z87.1
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit