Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-Verapamil hydrochloride, 99+%

Click to view available options
Quantity:
1g
152-11-4
C27H39ClN2O4
491.069
MFCD00055208
DOQPXTMNIUCOSY-UHFFFAOYSA-N
verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
62969
2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl
152-11-4
C27H39ClN2O4
491.069
MFCD00055208
DOQPXTMNIUCOSY-UHFFFAOYSA-N
verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
62969
2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

A calcium channel blocker with a chemopreventive character

Applications
±)-Verapamil hydrochloride is a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. It is also a calcium channel-blocker. It acts by inhibiting the slow channel e
Melting Point 138°C to 141°C
Color White
Quantity 1g
UN Number UN2811
Beilstein 3647093
Merck Index 14,9950
Solubility Information Soluble in water; Also soluble in ethanol or methanol
Formula Weight 491.07
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material (+/-)-Verapamil hydrochloride
Show More Show Less

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-800-5

RTECSNumber : YV8320000

TSCA : No

Recommended Storage : Ambient temperatures