Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Veratrole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
It is useful for organic synthesis of aromatic compounds. Veratrole is relatively electron-rich and thus readily undergoes electrophilic substitution.
Solubility
Soluble in alcohol, diethyl ether, acetone, and methanol. Slightly soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 21°C to 23°C |
Density | 1.084 |
Boiling Point | 206°C to 207°C |
Flash Point | 87°C (188°F) |
Refractive Index | 1.533 |
Quantity | 1000 g |
Beilstein | 1364621 |
Merck Index | 14,9956 |
Solubility Information | Soluble in alcohol,diethyl ether,acetone,and methanol. Slightly soluble in water. |
Formula Weight | 138.17 |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332 |
Hazard Statement | GHS H Statement H302-H227 Harmful if swallowed. Combustible liquid. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
EINECSNumber | 202-045-3 |
RTECSNumber | CZ6475000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction