Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Aluminum (Al) Standard for AAS 1000 ppm in 2% HCl, Ricca Chemical
SDP

Catalog No. p-7015018
Click to view available options
Quantity:
100 mL
500 mL

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Chemical Identifiers

CAS 7784-27-2, 7647-01-0, 7446-70-0, 7732-18-5
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 0.955598, 0.01%, 0.004802
CAS Max % 99.4602, 0.0204, 0.004998
Specific Gravity 1.013
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Industry Type Atomic Absorption
Quantity 100 mL
Grade High Purity
Chemical Name or Material Aluminum (Al) Standard for AAS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.