Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Antimony 1000 ppm in 20% HCl, Ricca Chemical
SDP

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Specifications

Packaging Natural LDPE Bottle
Specific Gravity 1
UN Number UN1789
Physical Form Liquid
CAS 7732-18-5, 7647-01-0, 10025-91-9
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RV010310100
SDP
View Documents
Ricca Chemical Company
RV010310100N
100 mL
Each for $108.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
RV010310250
SDP
View Documents
Ricca Chemical Company
RV010310250N
250 mL
Each for $188.90
N/A
Due to product restrictions, please to purchase or view availability for this product.
RV010310500
SDP
View Documents
Ricca Chemical Company
RV010310500N
500 mL
Each for $287.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • 1000 ppm in 20% HCl
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Natural LDPE Bottle
UN1789
7732-18-5, 7647-01-0, 10025-91-9
81.4062%, 20.4%, 0.1938%
1
Liquid
78.2138%, 19.6%, 0.1862%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit