Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Cesium Standard for ICP, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical

Click to view available options
Quantity:
50mL
100mL
250mL
500mL
7647-17-8, 7732-18-5
ClCs
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
7647-17-8, 7732-18-5
ClCs
168.355
AIYUHDOJVYHVIT-UHFFFAOYSA-M
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]

Formulated for high performance and reliable results

  • 1000ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
CAS Min % 97.8726%, 0.1274%
CAS Max % 100%, 0.1326%
Physical Form Liquid
Grade High Purity
Packaging Bottle, Poly Natural
Quantity 50mL
Specific Gravity 1