Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Interference Check, Analytes B; ISO 17025/Guide 34, Ricca Chemical

Formulated for high performance and reliable results

Specifications

Packaging Bottle, Poly Natural
Specific Gravity 1.08
UN Number UN3264
Physical Form Liquid
CAS Min % 92.6492%, 4.9%, 0.196%, 0.0686%, 0.0294%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RV010792500
View Documents Promotion Details
Ricca Chemical Company
RV010792500N
500mL
Each for $840.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
RV010792100
View Documents Promotion Details
Ricca Chemical Company
RV010792100N
100mL
Each for $412.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description

Description

  • 100ppm: Cd, Ni, Zn, 60ppm Sb; 50ppm Ba, Be, Co, Cr, Cu, Mn, V; 20ppm Ag; 10ppm As, Tl; 5ppm Pb, Se
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Specifications

Specifications

Bottle, Poly Natural
UN3264
92.6492%, 4.9%, 0.196%, 0.0686%, 0.0294%
7697-37-2, 7664-39-3, 13597-99-4, 7779-88-6
1.08
Liquid
96.4308%, 5.1%, 0.204%, 0.0714%, 0.0306%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit