Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Nexion ICP-MS Calibration Standard for Set Up 8, Ricca Chemical
1 ppb: Be, Ce, Fe, In, Li, Mg, Pb, U /nManufactured and Tested in an ISO 17025/Guide 34 Facility
Supplier: Ricca Chemical Company RV010880100N
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Natural LDPE Bottle | |
4.9%, 1.96%, 0.9114% | |
7790-69-4, 7697-37-2, 7647-01-0, 7732-18-5 | |
High Purity | |
Liquid |
100 mL | |
5.1%, 2.04%, 0.9486% | |
UN3264 | |
1.06 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction