Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Propionate - C2H5COO- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
SDP

Catalog Number p-7074089
Click to view available options
Quantity:
100 mL
250 mL
500 mL
137-40-6, 7732-18-5
C3H5NaO2
96.061
JXKPEJDQGNYQSM-UHFFFAOYSA-M
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]
137-40-6, 7732-18-5
C3H5NaO2
96.061
JXKPEJDQGNYQSM-UHFFFAOYSA-M
2723816
sodium;propanoate
CCC(=O)[O-].[Na+]

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis
Physical Form Liquid
CAS Min % 97.8726%, 0.1274%
CAS Max % 100%, 0.1326%
Grade High Purity
Packaging Natural LDPE Bottle
Quantity 100 mL
Specific Gravity 1.01
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit