Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Rubidium Standard for ICP 1000 ppm in H2O, Ricca Chemical
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Packaging | Natural LDPE Bottle |
Quantity | 100 mL |
CAS Min % | 97.8628% |
CAS Max % | 100% |
Grade | High Purity |
Specific Gravity | 1 |
Physical Form | Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction